Skip to Content
MilliporeSigma

96836

Acetic acid 100%

tested for PFAS methods, LiChropur™

Sustainability features

Synonym(s):

Acetic acid, Glacial acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein/REAXYS Number:
506007
MDL number:
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

suitable for EPA 1633, suitable for EPA 533, suitable for EPA 537.1

vapor density

2.07 (vs air)

assay

≥99.8% (acidimetric)

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F, 4 %, 59 °F

sustainability

Sustainability features

technique(s)

HPLC: suitable, LC/MS: suitable, ion chromatography: suitable

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

cation traces

Al: ≤5.0 ppb, Ca: ≤20.0 ppb, Fe: ≤20.0 ppb, K: ≤ 2.0 ppb, Mg: ≤2.0 ppb, Na: ≤5.0 ppb

greener alternative category

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

General description

We are committed to providing greener alternatives that adhere to the 12 Principles of Greener Chemistry. This acetic acid is derived from sustainably sourced beechwood, a renewable biomass resource, helping to reduce reliance on fossil-derived raw materials and supporting efficient use of natural resources. Consequently, it aligns with the Green Chemistry principles of "Use of Renewable Feedstocks" and "Waste Prevention."

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany


Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

Flash Point (°F)

102.2 °F - closed cup

Flash Point (°C)

39 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library