Skip to Content
MilliporeSigma

97072

Extractables and Leachables Silicone Mix

12 components in TBME, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

chemicals and industrial polymers
cosmetics
food and beverages

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.

Certified content incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com.

Check out our complete range of extractables and leachables certified reference mixes
The mix contains the following components in 50mg/L nominal concentration in tert-Butyl methyl ether (for exact concentration please check the Certificate):

  • Octamethylcyclotetrasiloxane 556-67-2
  • Dodecamethylcyclohexasiloxane 540-97-6
  • 1,1,3,3,5,5,7,7,9,9-Decamethylpentasiloxane 995-83-5
  • 1,1,3,3,5,5,7,7,9,9,11,11,13,13,15,15-Hexadecamethyloctasiloxane 19095-24-0
  • 1-Cyclohexylethanone 823-76-7
  • Cyclohexanol 108-93-0
  • Decamethylcyclopentasiloxane 541-02-6
  • Dodecamethylpentasiloxane 141-63-9
  • Tetradecamethylhexasiloxane 107-52-8
  • [2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]trimethylsilane 864079-62-9
  • Bis(2-ethylhexyl) adipate (certified value is nonstereo specific) 103-23-1
  • Di-n-octyl phthalate 117-84-0

Application

The certified reference material (CRM) is intended to be used as a calibrant for Silicone Extractables and Leachables testing or the testing of Food Contactmaterials (FCM). It contains the most commonly found Extractables from Silicone materials to help with identification and quantification.

Features and Benefits

  • Produced and certified under ISO/IEC 17025 and ISO 17034 accreditation
  • Traceable to certified NIST SRMs
  • Accompanied with a comprehensive Certificate of Analysis (CoA) (up to four pages), following ISO 33401:2024 (formerly ISO Guide 31), with data on stability, density, homogeneity, the accuracy of concentration, uncertainty, and traceability
  • Gravimetrically prepared using high-quality qNMR-characterized starting materials and qualified precision balances to ensure minimal uncertainty
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Available in well-sealed amber glass ampules

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

Flash Point (°F)

-18.4 °F - closed cup

Flash Point (°C)

-28 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library