Skip to Content
MilliporeSigma

97079

Brevetoxin 2 solution

15 μg/g in acetonitrile (nominal concentration), analytical standard

Synonym(s):

Brevetoxin B solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H70O14
CAS Number:
Molecular Weight:
895.08
NACRES:
NA.21
MDL number:
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

concentration

15 μg/g in acetonitrile (nominal concentration)

technique(s)

HPLC: suitable, LC/MS: suitable, liquid chromatography (LC): suitable, mass spectrometry (MS): suitable

density

0.786 g/mL

application(s)

food and beverages

format

single component solution

storage temp.

−20°C

InChI

1S/C50H70O14/c1-25(24-51)14-28-17-37(52)50(8)41(54-28)19-33-34(61-50)18-32-29(55-33)10-9-12-46(4)42(58-32)23-49(7)40(62-46)21-39-47(5,64-49)13-11-30-44(60-39)26(2)15-31-36(56-30)22-48(6)38(57-31)20-35-45(63-48)27(3)16-43(53)59-35/h9-10,16,24,26,28-42,44-45,52H,1,11-15,17-23H2,2-8H3/b10-9-/t26-,28-,29-,30+,31+,32+,33+,34-,35+,36-,37+,38-,39+,40-,41-,42-,44-,45-,46+,47-,48+,49+,50+/m1/s1

InChI key

LYTCVQQGCSNFJU-FGRVLNGBSA-N



Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

35.6 °F - closed cup

Flash Point (°C)

2.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library