Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
certified reference material
agency
ERM®
manufacturer/tradename
JRC
storage temp.
2-8°C
Analysis Note
A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage
please visit the JRC webpage
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants:
Pentachlorobenzene
α-Hexachlorocyclohexane
β-Hexachlorocyclohexane
γ-Hexachlorocyclohexane
Indicative Value:
δ-Hexachlorocyclohexane
Additional Material Information:
Extractable Fat
Matrix Group: Fish and Seafood
Organic Pollutants:
Pentachlorobenzene
α-Hexachlorocyclohexane
β-Hexachlorocyclohexane
γ-Hexachlorocyclohexane
Indicative Value:
δ-Hexachlorocyclohexane
Additional Material Information:
Extractable Fat
Matrix Group: Fish and Seafood
Legal Information
ERM is a registered trademark of European Commission
Storage Class
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.