Skip to Content
MilliporeSigma

SP2-PF45

Supelpak™-2 4PFAS 20-40 mesh

pkg of 500 g, XAD®-2 PS-DVB resin

Sustainability features

Synonym(s):

PS-DVB resin, XAD®-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
Packaging:
pkg of 500 g
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

suitable for EPA OTM-45, suitable for EPA OTM-50

packaging

pkg of 500 g

sustainability

Sustainability features

matrix

polymeric styrene/divinylbenzene

greener alternative category

General description

Amberlite™ XAD-2 resin is and has been crucial for collecting air contaminants. This polymeric adsorbent captures volatile organic compounds in air samples, enabling accurate assessments of environmental pollutants. However, its effectiveness relies heavily on a thorough cleaning process, which can be complex and involves multiple solvent washes and drying phases to eliminate residual contaminants that could compromise analytical results.
We are committed to bringing you Greener AlternativeProducts. This product is a Design for Sustainability (DfS) developed product,which belongs to one of the four categories of greener alternatives. Click hereto view its DfS scorecard.

Application

Interlaboratory Study for PFAS Background Extracted from Multiple Pre-cleaned XAD-2 Adsorbent Lots - View poster

Analysis Note

Supelpak™-2 4PFAS is designed specifically for EPA Other Test Method 45 (OTM-45): Measurement of Selected Per- and Polyfluorinated Alkyl Substances from Stationary Sources. This resin has been rigorously tested for background PFAS levels (CoA is listed with testing levels for the analytes within OTM-45). Integrating the ready-to-use Supelpak™-2 4PFAS into your air sampling protocols will streamline processes and enhance the accuracy of PFAS analyses. View EPA method- Background cleanliness and recovery of Isotope Dilution Standards.


Optimizing Your OTM-45 Workflow webinar link: Do′s and Don′ts from a Panel of PFAS Experts.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Supelpak is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Disclaimer

PFAS Cross-Contamination Prevention Notice
This product has been tested for per- and polyfluoroalkyl substances (PFAS). Users should pay special attention to avoid cross-contamination due to the prevalenceof PFAS related chemicals in consumer products, equipment, and in the environment.
Some best practices to maintain sample integrity and prevent cross-contamination:
  • Use only PFAS-free materials when handling this product (recommended: stainless steel, silicone, high-density polyethylene)
  • Avoid contact with potential PFAS sources including PTFE materials, waterproof clothing, food packaging, and certain personal care products
  • Wear nitrile gloves and wash hands thoroughly before handling Implement appropriate field and laboratory blanks to verify absence of contamination
  • Follow all method-specific quality control requirements for sampling, preservation, and analysis


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library