Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C20H28O10
CAS Number:
Molecular Weight:
428.43
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
eCl@ss:
25140904
grade
pharmaceutical primary standard
agency
BP
API family
rosavin
form
solid
manufacturer/tradename
EDQM
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
O[C@H]1CO[C@@H](OC[C@H]2O[C@@H](OC\C=C\c3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H]1O
InChI
1S/C20H28O10/c21-12-9-28-19(17(25)14(12)22)29-10-13-15(23)16(24)18(26)20(30-13)27-8-4-7-11-5-2-1-3-6-11/h1-7,12-26H,8-10H2/b7-4+/t12-,13+,14-,15+,16-,17+,18+,19-,20+/m0/s1
InChI key
RINHYCZCUGCZAJ-IPXOVKFZSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Biochem/physiol Actions
Rosavin is a well-known glycoside antioxidant from Rhadiola rosea (Golden Root, or Arctic Root) that exhibits antidepressant, anxiolytic and adaptogenic properties.
Rosavin is an antidepressant and anxiolytic; antiapoptotic.
Packaging
Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.