Select a Size
About This Item
grade
pharmaceutical primary standard
agency
USP
form
powder
manufacturer/tradename
USP
application(s)
USP Biologics
pharmaceutical
format
neat
storage temp.
−20°C
SMILES string
P(N(C(C)C)C(C)C)(O[C@@H]1[C@H](O[C@H](C1)[n]5c6nc([nH][c](c6nc5)=O)NC(=O)C(C)C)COC(c4ccc(cc4)OC)(c3ccc(cc3)OC)c2ccccc2)OCCC#N
InChI
1S/C44H54N7O8P/c1-28(2)41(52)48-43-47-40-39(42(53)49-43)46-27-50(40)38-25-36(59-60(57-24-12-23-45)51(29(3)4)30(5)6)37(58-38)26-56-44(31-13-10-9-11-14-31,32-15-19-34(54-7)20-16-32)33-17-21-35(55-8)22-18-33/h9-11,13-22,27-30,36-38H,12,24-26H2,1-8H3,(H2,47,4
InChI key
FDRMKYVTIFSDPR-MMROLVBFSA-N
General description
This reagent is an isobutyryl-protected deoxyguanosine phosphoramidite, used for the incorporation of deoxyguanosine residues into synthetic DNA strands.
The USP biologics oligonucleotides category includes a variety of synthetic nucleic acid sequences that are essential in research, diagnostics, and therapeutic applications. These oligonucleotides play crucial roles in molecular biology, including gene expression regulation, antisense therapy, and RNA interference. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of oligonucleotide products throughout their lifecycle.
The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.
Analysis Note
Other Notes
Storage Class
13 - Non Combustible Solids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.