Skip to Content
MilliporeSigma

1156167

USP

3-Cyclopentylpropionic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cyclopentanepropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H9CH2CH2CO2H
CAS Number:
Molecular Weight:
142.20
MDL number:
NACRES:
NA.21
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sodium cyclamate

manufacturer/tradename

USP

refractive index

n20/D 1.457 (lit.)

bp

130-132 °C/12 mmHg (lit.)

density

0.996 g/mL at 25 °C (lit.)

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CCC1CCCC1

InChI

1S/C8H14O2/c9-8(10)6-5-7-3-1-2-4-7/h7H,1-6H2,(H,9,10)

InChI key

ZRPLANDPDWYOMZ-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Still not finding the right product?

Explore all of our products under 3-Cyclopentylpropionic acid


Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

3 - Flammable liquids

WGK

WGK 3

Flash Point (°F)

116.6 °F - closed cup

Flash Point (°C)

47 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library