Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
pharmaceutical primary standard
agency
USP
form
liquid
manufacturer/tradename
USP
packaging
pkg of (2 x 100 mg)
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
O(C)C1COC2C(OC)COC12
InChI
InChI=1S/C8H14O4/c1-9-5-3-11-8-6(10-2)4-12-7(5)8/h5-8H,3-4H2,1-2H3/t5-,6+,7-,8-/m1/s1
InChI key
MEJYDZQQVZJMPP-ULAWRXDQSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Still not finding the right product?
Explore all of our products under Dimethyl Isosorbide
Storage Class
10 - Combustible liquids
Flash Point (°F)
240.8 °F - closed cup
Flash Point (°C)
116 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.