Skip to Content
MilliporeSigma

1256037

USP

Ethacrynic Acid Related Compound C

Synonym(s):

({4-[2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl]-2,3- dichlorophenoxy}acetic acid)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H24Cl4O8
CAS Number:
Molecular Weight:
606.28
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

format

neat

storage temp.

2-30°C

SMILES string

O=C(O)COC1=CC=C(C(=O)C2(OC(C=3C=CC(OCC(=O)O)=C(Cl)C3Cl)=C(CC)CC2)CC)C(Cl)=C1Cl

InChI key

InChI=1S/C26H24Cl4O8/c1-3-13-9-10-26(4-2,25(35)15-6-8-17(23(30)21(15)28)37-12-19(33)34)38-24(13)14-5-7-16(22(29)20(14)27)36-11-18(31)32/h5-8H,3-4,9-12H2,1-2H3,(H,31,32)(H,33,34)

General description

Find answers to frequently asked questions on USP products in USP FAQs.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library