Skip to Content
MilliporeSigma

1269383

USP

Febuxostat Related Compound C

Synonym(s):

Ethyl 2-(3-cyano-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H12N2O3S
CAS Number:
Molecular Weight:
288.32
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

storage temp.

2-8°C

SMILES string

N#CC=1C=C(C=CC1O)C2=NC(=C(S2)C(=O)OCC)C

InChI key

InChI=1S/C14H12N2O3S/c1-3-19-14(18)12-8(2)16-13(20-12)9-4-5-11(17)10(6-9)7-15/h4-6,17H,3H2,1-2H3

General description

Find answers to frequently asked questions on USP products in USP FAQs.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library