Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(C19H24N2)2 · C23H16O6
CAS Number:
Molecular Weight:
949.18
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
grade
pharmaceutical primary standard
agency
USP
API family
imipramine
manufacturer/tradename
USP
application(s)
pharmaceutical
pharmaceutical (small molecule)
SMILES string
N8(c9c(cccc9)CCc%10c8cccc%10)CCCN(C)C.N5(c6c(cccc6)CCc7c5cccc7)CCCN(C)C.Oc1c(c4c(cc1C(=O)O)cccc4)Cc2c3c(cc(c2O)C(=O)O)cccc3
InChI
1S/C23H16O6.2C19H24N2/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;2*1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2*3-6,8-11H,7,12-15H2,1-2H3
InChI key
BPQZYOJIXDMZSX-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
