Skip to Content
MilliporeSigma

1351016

USP

Isoproterenol Related Compound A

Synonym(s):

(1-(3,4-Dihydroxyphenyl)-2-(isopropylamino)ethan-1-one hydrochloride)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H15NO3.ClH
CAS Number:
Molecular Weight:
245.70
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

No data available

Flash Point (°C)

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library