Skip to Content
MilliporeSigma

1357089

USP

Teriflunomide Related Compound B

Synonym(s):

2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H7F3N2O
Molecular Weight:
228.17
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library