Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C3H8N4O2
Molecular Weight:
132.12
NACRES:
NA.24
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
agency
USP
manufacturer/tradename
USP
format
neat
storage temp.
2-30°C
General description
Find answers to frequently asked questions on USP products in USP FAQs.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
No data available
Flash Point (°C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
