Skip to Content
MilliporeSigma

1445480

USP

Monascin

Synonym(s):

(35,3aR,9aR)-3-Hexanoyl-9a-methyl-6-((E)-prop-1-en-1-yl)-3a,4,8,9a-tetrahydro-2H- furo[3,2-g]isochromene-2,9(3H)-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

format

neat

storage temp.

-10 to -25°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.


Still not finding the right product?

Explore all of our products under Monascin


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library