Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H10N2O3
CAS Number:
Molecular Weight:
158.16
NACRES:
NA.24
UNSPSC Code:
41116107
grade
USP Analytical Reference Materials (ARMs)
agency
USP
manufacturer/tradename
USP
format
neat
storage temp.
2-8°C
General description
Find answers to frequently asked questions on USP products in USP FAQs.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.