Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H7FO
CAS Number:
Molecular Weight:
126.13
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
agency
USP
form
powder
shelf life
see USP product information sheet
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
USP issued SDS can be found here
Storage Class
10 - Combustible liquids
Flash Point (°F)
194.0 °F
Flash Point (°C)
90.0 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.