Skip to Content
MilliporeSigma

1724907

USP

Zolpidem tartrate

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H42N6O2 · C4H6O6
CAS Number:
Molecular Weight:
764.87
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


API family

zolpidem

manufacturer/tradename

USP

drug control

kontrollierte Droge in Deutschland, psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

application(s)

pharmaceutical (small molecule)

SMILES string

[n]21c(nc(c2CC(=O)N(C)C)c3ccc(cc3)C)ccc(c1)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O

InChI

1S/C19H21N3O.C4H6O6/c1-13-5-8-15(9-6-13)19-16(11-18(23)21(3)4)22-12-14(2)7-10-17(22)20-19;5-1(3(7)8)2(6)4(9)10/h5-10,12H,11H2,1-4H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1

InChI key

NYVVVBWEVRSKIU-LREBCSMRSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


Still not finding the right product?

Explore all of our products under Zolpidem tartrate


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library