Select a Size
About This Item
grade
USP Analytical Reference Materials (ARMs)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
-10 to -25°C
SMILES string
O=C(O)CCC(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C1NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(N)CSSC1)CO)CC(=O)N)CC(C)C)CO)C(O)C)C(C)C)CC(C)C)CCCCN)CC(C)C)CO)CCC(=O)N)C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)
InChI
InChI=1S/C145H239N43O49S2/c1-65(2)45-86(174-139(232)110(70(11)12)182-136(229)99-63-239-238-62-79(148)117(210)177-96(59-190)134(227)173-92(52-103(150)200)131(224)171-90(49-69(9)10)129(222)179-98(61-192)135(228)186-114(74(16)196)142(235)180-99)118(211)157-5
InChI key
VDUREYOUDGHTGP-VJVYQDLKSA-N
General description
Analysis Note
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.