Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H20ClN3O4S
CAS Number:
Molecular Weight:
409.89
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
grade
pharmaceutical analytical impurity (PAI)
agency
USP
API family
rivaroxaban
manufacturer/tradename
USP
application(s)
pharmaceutical
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
-10 to -25°C
SMILES string
[s]1c(ccc1C(=O)NC[C@H](O)CNc2ccc(cc2)N3CCOCC3=O)Cl
InChI
1S/C18H20ClN3O4S/c19-16-6-5-15(27-16)18(25)21-10-14(23)9-20-12-1-3-13(4-2-12)22-7-8-26-11-17(22)24/h1-6,14,20,23H,7-11H2,(H,21,25)/t14-/m1/s1
InChI key
OKMRQXXUAFSBCM-CQSZACIVSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
Rivaroxaban Open Ring is a USP Pharmaceutical Analytical Impurity (PAI).
USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.
Associated Drug Substance: Rivaroxaban
Therapeutic Area: Anticoagulants & Antiplatelets
USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.
Associated Drug Substance: Rivaroxaban
Therapeutic Area: Anticoagulants & Antiplatelets
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.