Skip to Content
MilliporeSigma

1A01240

USP

(16a)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

(16α)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione, 2-((8S,9S,10R,13S,14S,16R,17S)-16,17-Dihydroxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,10,11,12,13, 14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl acetate, (2-((8S,9S,10R,13S,14S,16R,17S)-16,17-Dihydroxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl acetate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H28O7
CAS Number:
Molecular Weight:
416.46
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

API family

prednisolone

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
(16α)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione is a USP Pharmaceutical Analytical Impurity (PAI).

USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

Associated Drug Substance: Budesonide

Therapeutic Area: Steroids

For more information about this PAI, visit here.

Application

(16α)-21-(Acetyloxy)-16,17-dihydroxypregna-1,4-diene-3,11,20-trione (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:

1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.


Still not finding the right product?


Pictograms

Health hazard

Signal Word

Warning

Hazard Codes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library