Skip to Content
MilliporeSigma

1A02940

USP

Ivabradine Amine Hydrochloride

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

Ivabradine Amine Hydrochloride, (S)-1-(3,4-Dimethoxybicyclo(4.2.0)octa-1,3,5-trien-7-yl)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H18ClNO2
CAS Number:
Molecular Weight:
243.73
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

API family

ivabradine

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

-10 to -25°C

SMILES string

Cl.N(C[C@H]1Cc2c1cc(c(c2)OC)OC)C

InChI

1S/C12H17NO2.ClH/c1-13-7-9-4-8-5-11(14-2)12(15-3)6-10(8)9;/h5-6,9,13H,4,7H2,1-3H3;1H/t9-;/m1./s1

InChI key

SWSAIQSQSDOONK-SBSPUUFOSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Ivabradine Amine Hydrochloride is a USP Pharmaceutical Analytical Impurity (PAI).


USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.


Associated Drug Substance: Ivabradine Hydrochloride


Therapeutic Area: Cardiovascular

Application

Ivabradine Amine Hydrochloride (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library