Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C8H13N2O7P · 2Na
Molecular Weight:
326.15
UNSPSC Code:
41116107
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
[Na].O=C1C=CN(C(=O)N1)CC(OCP(=O)(O)O)CO
InChI
InChI=1S/C8H13N2O7P.2Na/c11-4-6(17-5-18(14,15)16)3-10-2-1-7(12)9-8(10)13;;/h1-2,6,11H,3-5H2,(H,9,12,13)(H2,14,15,16);;/t6-;;/m0../s1
InChI key
PRZWYPAUYCTWBZ-ILKKLZGPSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.