Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C24H33N3O5
CAS Number:
Molecular Weight:
443.54
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
powder
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Pharmaceutical QC release and stability testing. Identify and quantify Related Compound A in amisulpride drug substance and drug product.
- Manufacturing and process control. Monitor impurity levels from raw material through finished product.
- System suitability and peak identification. Confirm the position of the impurity peak and support chromatographic performance checks.
- Analytical method development and validation. Establish specificity, linearity, limits of detection and quantitation.
Features and Benefits
- Official USP Reference Standard. Provided as delivered and specified by the United States Pharmacopeia, and accompanied by documentation issued under USP′s authority.
- Backed by a trusted supply chain. It is available from MilliporeSigma with the global distribution and customer support, alongside a wide portfolio of USP standards.
- Confidence in compliance. A pharmacopeial standard supports your regulatory submissions and quality documentation, so your results rest on an accepted reference.
- Accurate impurity identification. An authentic reference lets you confirm retention time and identity, which reduces misidentification risk during method development and routine testing.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
USP issued SDS can be found here
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT RE 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

