Skip to Content
MilliporeSigma

1A18400

USP

N-Methyl Moxifloxacin

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

N-Methyl Moxifloxacin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H26FN3O4
Molecular Weight:
415.46
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

form

powder

shelf life

per USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the issuing Pharmacopoeia.">website

Application

  • Pharmaceutical QC release and stability testing. Identify and quantify Related Compound A in amisulpride drug substance and drug product.
  • Manufacturing and process control. Monitor impurity levels from raw material through finished product.
  • System suitability and peak identification. Confirm the position of the impurity peak and support chromatographic performance checks.
  • Analytical method development and validation. Establish specificity, linearity, limits of detection and quantitation.

Features and Benefits

  • Official USP Reference Standard. Provided as delivered and specified by the United States Pharmacopeia, and accompanied by documentation issued under USP′s authority.
  • Backed by a trusted supply chain. It is available from MilliporeSigma with the global distribution and customer support, alongside a wide portfolio of USP standards.
  • Confidence in compliance. A pharmacopeial standard supports your regulatory submissions and quality documentation, so your results rest on an accepted reference.
  • Accurate impurity identification. An authentic reference lets you confirm retention time and identity, which reduces misidentification risk during method development and routine testing.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here">here


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library