Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
ASTM D3612-96: GC Analysis of Transformer Gas on Carboxen®-1010 PLOT
ASTM D3612-96: GC Analysis of Transformer Gas on Carboxen®-1010 PLOT
Conditions
column Carboxen-1010 PLOT, 30 m x 0.53 mm I.D. (25467)
oven 35 C (8.5 min), 24 °C/min to 250 °C
carrier gas argon, 3.0 mL/min
sample 9 analytes, each at 500 ppm
injection 10.0 µLdirect using a sample valve, 150 °C
inj. temp. 200 °C
detector TCD, 230 °C, Methanizer, 375 C, and FID, 230 °C in series
Description
Categories Analytical Chromatography, GC Applications, Light Hydrocarbons, Gases, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • Acetylene
  • Carbon dioxide
  • Carbon monoxide
  • Ethane
  • Ethylene
  • Hydrogen
  • Methane
  • Nitrogen
  • Oxygen