Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | $316.00 |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | $568.00 |
About This Item
$316.00
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GACCGTTTTGGAAGCATTGTTCTTTTCACTGTTAGTGAGGAGCGGTCAAGGATGCGTATTAAGAAAATAACTAGAAGTGGCCGCATGATGTACCTCTCACCAATGAGATGCCTGTCAAGGGTATACGTGGGTCTTTTGAGATCTTATTCACTGATTTGAAGAGTTCATGCATCGAGCCAGTAGGCTGTGCACATGGGTAAAAACTGGCTGCGGCTAAACAGTTACTCTTAGAAGAAAGGAAGCATTGAGAGTAAGGAACAAATGCCAATGATGACAACCCAAACACTATGTATTGTAAATGAAGTGGACTTCGTGGTTTCTTTATCCACCAATAAATTAAAAGAAAACTTACAGGAAATGGACACAGCCCTGGATAAATGC
GenBank accession no.
shipped in
ambient
storage temp.
−20°C
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting human CTD-2001C12.1 | description targeting human RP1-309F20.3 | description targeting mouse Ak085217 | description targeting human AB015752.3 |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| esiRNA cDNA target sequence GACCGTTTTGGAAGCATTGTTCTTTTCACTGTTAGTGAGGAGCGGTCAAGGATGCGTATTAAGAAAATAACTAGAAGTGGCCGCATGATGTACCTCTCACCAATGAGATGCCTGTCAAGGGTATACGTGGGTCTTTTGAGATCTTATTCACTGATTTGAAGAGTTCATGCATCGAGCCAGTAGGCTGTGCACATGGGTAAAAACTGGCTGCGGCTAAACAGTTACTCTTAGAAGAAAGGAAGCATTGAGAGTAAGGAACAAATGCCAATGATGACAACCCAAACACTATGTATTGTAAATGAAGTGGACTTCGTGGTTTCTTTATCCACCAATAAATTAAAAGAAAACTTACAGGAAATGGACACAGCCCTGGATAAATGC | esiRNA cDNA target sequence TTCAACTGGACCACCAGCTACTACAACCCTAGAAAAATGGAGGCACAGATATGTGGGTCCCAGAGCCCTGGGGGAGCTGGAATGGATACTCGCTAACACACGGGACACGAGATTCTCTTTCCTTTGGAAGGGTCCTGTCCATGGCCCTTCTTGGGTTCTGGACTGTCCTACAATGCCTGACTTTAGTTTCTGTGGATCATGATAAGCATTATTCATTTTTTCCCCCTCAAATATCCCACAGCTTCAAACTTACCTGAATGCAGGTTCCCTGTAAAAATACCATCATGTAATTTGCACATTAGCAATCAGTTTGATGAATGGCAAAATCGTTCAAATGGAAAAGAGGAGAGAGATAGTGTGGTCTTTTGATGGAGGGGGAGAGGCAAGAGCCAGATAAATCAGAAGAGAGAGGCGAGGCTCTTGCAGCCCAAGTCTTAATTTCCTTGTAAGAATCGATGCTCACTCAGTGGTGACACTGAGCTTGTGAAAGCTCAACTGTGACACTCTTTAGAAAGTAGTACAGAGATTTTTTTCCTTCCCATTATCAACTCA | esiRNA cDNA target sequence CTGGATCTGCACCTGAGACATGAATTCAAGTACCAACTGCAGGTCTCAGCAGCCTAGGAATCTCAGACAAGTACTTTATCTTCTGTTGATATCTCAGAGTTGCTGTGAGTATCAAGTACAATACTTACTCCTCAGCATACCTGTTACCCAGCACTTAGCAAATGAGAGCAGAATTCTTTAAAGTCTGTAGTGCTAGATATGGAACTTGAGACCTGTGCATCTAGGCAAGCTCTCTACTACAGAGCCCCATTCCCCCACCCCTCAGTTTGTTAAGATAGGATTTCACTATGAAGTCTTGGCTGGCTTCAAACTCAAGATCTTCTTGCCTCTACCTCCTGTATGCTGGGATCACA | esiRNA cDNA target sequence TCCTGTGTCATGCTCCACATTCTGTCCATTCATCTCTCAGTTAACTAAATAGAAGTGATGCAAGGGAGGACTCCAAGGAAGCCCTAGGGATCATGAAGCTACAGAATGGGAGAATTGCTGTCACCACAGTTCTGGAAGAAGAATTGTTCAGGAGAACAGCCATTGGACTATGGCATGCCAAAGAAATCTCTGTTGTGAAACCACAATTAAAAGTTGTTCTTCCTGCCTCTGCTTCTTGACCAACC |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| GenBank accession no. | GenBank accession no. | GenBank accession no. | GenBank accession no. |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
