Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | $316.00 |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | $568.00 |
About This Item
NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207
Sign in to see your pricing
$316.00
PRICE WITHOUT NATIONAL TAXES
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AGTCGAGAGCTGGAGTCAGGCGCCGCTCACGCTGGGATGACGGAGTGTTAACCTCAGACTCTGGGTCCCTTTGGGCCGAAGCTTTAAGAAAACAGGGGCAGCGAGCCCGGCGTCCCAAGGAGCCTTGCCAAACTGGGCATGGAGAACGGGCCTCCTGGATGGCCACCCGCCCAGCTGCCAAAGAGGCCGGAGAGCTGGAGCCGGGACTTGGAGGATCTTCTTCAGAGAGGCAGGAGGGCGGTCCGAACGTGCCCCCACCCCTGACCTCAAGCCCATGTGGAGAGGGCCAACAGAGCCTCTTGGAGAGCTGGATTTGCGTCCTCTGCAGCTTGGGGGACACGTCGTTGATGGTGACTGACACGCCCCAGAGATAGTGACATGATGATTAAGAGTGACAGCCTGAGGGA
Ensembl | human accession no.
shipped in
ambient
storage temp.
−20°C
General description
MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.