Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | $316.00 |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | $568.00 |
About This Item
$316.00
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GCTCGCCCTAGATAAACGTCAAAGGAGGCGGTCAGCGCTCTGAAAAGCTAAGCTTTTCCCGGCACATCCCTGTGTGCGAGCACATGCCAAGTACGTGAATGCAAACATATGTATTTAAGCTGGATTTATCATTCTGCACCTTAGCGTGAAGCCAGCTTCAGAACTTATATAAAAGTGAAAAGCGCCCTCAAAAGTTCAGACTTGCCCCCCCCATCCCCGGCGAAGTTCAAACTCTGCCTGGCTTTGCTCGCTCTTTCCCTACCAGCACTCAGACCGACGAGCTTGTTGGCTCAGAGAGATCCAGAAGCGGTTTCTAAGAACTAGTCACGTACAGTCTGGGCTCAGCAACGTGTCCAGCTGTTTTCTGCGCCTGTTACCCTGTTTGCACGATCCAGTTAACCCAGATGTTAAAGGACAGAGCCCAGAGAGAGCAGTGGAATGCGTGGAATGTACCCCTGGTGGACCTCTATGTAGCTTGACCTGATTGATGTCCTGGGAATCCCAGAATCCAAAGTTGG
shipped in
ambient
storage temp.
−20°C
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.