Skip to Content
Merck

EMU173011

MISSION® esiRNA

targeting mouse Fstl1

Sign In to View Organizational & Contract Pricing

Select a Size

20 μG

$287.00

50 μG

$517.00

$287.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCTGCTCCTGTTTCTAGGTGTCAGTTTACTTAGTAAACTGGTAAGAATGAATCTTGGAATTTAATAAATGGTAGTTTGTGGTTTAGCCAACTGGTCCAGAGGGAGCTACCTTCTCCTTAGGATAGATGAATCTACACCATAAGAAAAACCAGCCAGGAATAGCATGGATGGGTTTTGCTTTGGTTGAAATGATCCTAGCAGGTGACTGGGTATGAGGACTTCATGGTCACTCTGCCCAGGAAGAGAGCGTGAAGGACAACTAGCAGCTTCCTTAGGGATGGTACACATGAGTGTGATCTCTGGAGTCAGAGGTTTCCCCACACACATGATGATAAAACTTTTCAGATTTAGAGCGGTTAAAACTGGAGATCGAATCTGGATTGAGAATCAGCACTGGGGGAGAAACTGTTATTGAAAGTCAATCCTTTCTTTGAGCCTCCGAATAAACTATGGAGATTTTCCTGCATAGGAAAGTGTGGAATGTTGAGCTATTGAGATGGGAGTGGAATTCG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU078691EMU013961EMU210391
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Gene Information

mouse ... FSTL1(14314), Fstl1(14314)

Gene Information

mouse ... FST(14313), Fst(14313)

Gene Information

mouse ... FBLN1(14114), Fbln1(14114)

Gene Information

mouse ... FTL1(100862446), Ftl1(14325)

esiRNA cDNA target sequence

AGCTGCTCCTGTTTCTAGGTGTCAGTTTACTTAGTAAACTGGTAAGAATGAATCTTGGAATTTAATAAATGGTAGTTTGTGGTTTAGCCAACTGGTCCAGAGGGAGCTACCTTCTCCTTAGGATAGATGAATCTACACCATAAGAAAAACCAGCCAGGAATAGCATGGATGGGTTTTGCTTTGGTTGAAATGATCCTAGCAGGTGACTGGGTATGAGGACTTCATGGTCACTCTGCCCAGGAAGAGAGCGTGAAGGACAACTAGCAGCTTCCTTAGGGATGGTACACATGAGTGTGATCTCTGGAGTCAGAGGTTTCCCCACACACATGATGATAAAACTTTTCAGATTTAGAGCGGTTAAAACTGGAGATCGAATCTGGATTGAGAATCAGCACTGGGGGAGAAACTGTTATTGAAAGTCAATCCTTTCTTTGAGCCTCCGAATAAACTATGGAGATTTTCCTGCATAGGAAAGTGTGGAATGTTGAGCTATTGAGATGGGAGTGGAATTCG

esiRNA cDNA target sequence

CCACTTGTGTGGTGGATCAGACCAATAATGCCTACTGTGTGACCTGTAATCGGATTTGCCCAGAGCCCTCCTCTTCTGAACAGTACCTTTGTGGAAATGATGGAGTGACTTACTCCAGCGCCTGCCACCTGAGAAAGGCCACCTGCTTGCTGGGCAGATCCATTGGATTAGCCTATGAGGGAAAGTGTATCACAAAGTCCTGTGAAGATATCCAGTGTGGCGGCGGGAAAAAATGCCTATGGGATTCCAAGGTTGGCAGAGGTCGCTGCTCTCTCTGCGATGAGCTGTGTCCTGACAGTAAGTCGGATGAGCCGGTCTGTGCCAGTGACAATGCCACATACGCCAGCGAGTGTGCCATGAAGGAAGCTGCCTGCTCTTCTGGCGTGCTTCTTGAAGTGAAGCATTCTGGATCTTGCAACTCCATCTCGGAAGAAACGGAGGAAGAGGAGGAGGAGGA

esiRNA cDNA target sequence

GCTCTTGCTTTGTGGGCTACCAGCTGCAGTCGGATGGTGTCTCCTGCGAAGATATCAATGAATGCATCACAGGCAGCCATAACTGCCGGCTGGGAGAATCCTGCATCAATACAGTGGGCTCTTTCCGCTGCCAGCGGGACAGCAGCTGTGGGACTGGCTATGAGCTCACAGAGGATAATAACTGCAAAGATATTGACGAATGTGAGACTGGTATTCATAACTGCCCCCCCGATTTTATCTGTCAGAATACTCTGGGATCCTTCCGTTGCAGACCGAAGCTGCAGTGCAAGAGCGGCTTCATACAGGATGCTCTAGGCAACTGCATTGATATCAATGAGTGTTTAAGTATCAGTGCTCCATGCCCTGTGGGGCAGACATGCATCAATACAGAGGGCTCCTACACATGCCAGAAGAATGTGCCCAACT

esiRNA cDNA target sequence

CCATCTCAAGATGAATGGGGTAAAACCCAGGAGGCCATGGAAGCTGCCTTGGCCATGGAGAAGAACCTGAATCAGGCCCTCTTGGATCTGCATGCCCTGGGTTCTGCCCGCGCGGACCCTCATCTCTGTGACTTCCTGGAAAGCCACTATCTGGATAAGGAGGTGAAACTCATCAAGAAGATGGGCAACCATCTGACCAACCTCCGCAGGGTGGCGGGGCCACAACCAGCGCAGACTGGCGCGCCCCAGGGGTCTCTGGGCGAGTATCTCTTTGAGCGCCTCACTCTCAAGCACGACTAGGAGGCCTCTGTACCTTCCAAGGGGCTCCCCCCTCTGCTCTGCACCAGCCCGCCCTGGGACCTCCACCTGAATGAACCTCTCAAGCCACTAGGCAGCTTTGTAACCGCCCTGGAGCCTCTGTCAAGTCTTGGACCA

Ensembl | mouse accession no.

ENSMUSG00000022816

Ensembl | mouse accession no.

ENSMUSG00000021765

Ensembl | mouse accession no.

ENSMUSG00000006369

Ensembl | mouse accession no.

ENSMUSG00000050708

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service