Merck
  • Cathinone, an active principle of Catha edulis, accelerates oxidative stress in the limbic area of swiss albino mice.
Cathinone, an active principle of Catha edulis, accelerates oxidative stress in the limbic area of swiss albino mice.
Journal of ethnopharmacology (2014-08-26)
Mohammed M Safhi, Mohammad Firoz Alam, Sohail Hussain, Mohammed Abdul Hakeem Siddiqui, Gulrana Khuwaja, Ibrahim Abdu Jubran Khardali, Rashad Mohammed Al-Sanosi, Fakhrul Islam
ABSTRACT
Cathinone hydrochloride is an active principle of the khat plant (Catha edulis) that produces pleasurable and stimulating effects in khat chewers. To the best of our knowledge no data of cathinone on oxidative stress in limbic areas of mice is available. This is the first study of cathinone on oxidative stress in limbic areas of the brain in Swiss albino male mice. The animals were divided into four groups. Group-I was the control group and received vehicle, while groups-II to IV received (-)-cathinone hydrochloride (0.125, 0.25 and 0.5 mg/kg body wt., i.p.) once daily for 15 days. The level of lipid peroxidation (LPO) was elevated dose-dependently and was significant (p<0.05, p<0.01) with doses of 0.25 and 0.5mg/kg body wt. of cathinone as compared to control group. In contrast, the content of reduced glutathione (GSH) was decreased significantly (p<0.01, p<0.001) with doses of 0.25 and 0.5mg/kg body wt. of cathinone as compared to control group. The activity of antioxidant enzymes (GPx, GR, GST, CAT, and SOD) was also decreased dose-dependently: the decreased activity of GPx, GR, catalase and SOD was significant with doses of 0.25 and 0.5 mg of cathinone as compared to control group, while the activity of GST was decreased dose-dependently and was significant with 0.5mg of cathinone as compared to control group. The results indicate that the cathinone generated oxidative stress hampered antioxidant enzymes, glutathione and lipid peroxidation.
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)