Merck
  • Ameliorative action of curcumin in cisplatin-mediated hepatotoxicity: an in vivo study in Wistar rats.
Ameliorative action of curcumin in cisplatin-mediated hepatotoxicity: an in vivo study in Wistar rats.
Archives of medical research (2014-08-01)
Mohammad Waseem, Priyanka Pandey, Babita Tomar, Sheikh Raisuddin, Suhel Parvez
ABSTRACT
Cisplatin (CP) is regarded as a major anticancer drug against a wide spectrum of leukemia and other malignancies. The mode of action of CP on cancer cells is attributed to its property of releasing free radicals which, at the same time, has the potential to damage liver and kidney cells. The tissue-specific toxicity of CP to the kidneys is well documented. However, there is a paucity of literature on systemic toxicity and also the amelioration of such effect. The aim of the present study was to evaluate the damage caused by CP and its modulation by using the antioxidant capacity of curcumin (CMN, a natural polyphenolic compound) in Wistar rats. We evaluated the ameliorative effect of CMN (200 mg/kg body weight, b.w.) on the toxicity of CP in rat liver. Oxidative stress biomarkers, enzymatic and nonenzymatic antioxidants were evaluated for assessing the mitigatory potential of CMN against CP-induced toxicity. A single dose of CP (7 mg/kg b.w.) caused marked hepatic damage. CP caused a significant increase in lipid peroxidation (LPO) level and protein carbonyl (PC) content. Pretreatment of rat with CMN significantly restored the LPO levels and PC content. It also replenished the CP-induced modulatory effects on altered enzymatic and nonenzymatic antioxidants in liver. Our results clearly demonstrated that the role of oxidative stress is detrimental in systemic toxicity induced by CP and suggest a mitigatory effect of CMN on CP-induced hepatotoxicity in rat.
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,2-Dinitrobenzene, 97%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder