Saltar al contenido
Merck

ZRB3108

Anti-HSD3B1 Antibody, clone 2M19 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sinónimos:

3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I, 3-beta-HSD I, 3-beta-hydroxy-5-ene steroid dehydrogenase, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase (EC:1.1.1.145), 3-beta-hydroxysteroid 3-dehydrogenase (EC:1.1.1.270), Delta-5-3-ketosteroid isomerase

Iniciar sesión para ver los posibles precios especiales de su organización

Seleccione un Tamaño

En este momento no podemos mostrarle ni los precios ni la disponibilidad

Acerca de este artículo

UNSPSC Code:
12352203

Saltar a

Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2M19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 42.25 kDa
observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... HSD3B1(3283)

General description

3-beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 (UniProt: P14060; also known as 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type I (3-beta-HSD I), 3-beta-hydroxy-5-ene steroid dehydrogenase, 3-beta-hydroxy-Delta(5)-steroid dehydrogenase (EC:1.1.1.145), 3-beta-hydroxysteroid 3-dehydrogenase (EC:1.1.1.270), Delta-5-3-ketosteroid isomerase) is encoded by the HSD3B1 (also known as 3BH, HSDB3A) gene (Gene ID: 3283) in human. HSD3B1 is an essential enzyme involved in steroid biosynthesis, catalyzing the conversion of 5-steroid precursors into their 4-keto forms, which are critical intermediates in the production of progesterone, androgens, and estrogens. HSD3B1 is a member of the short-chain dehydrogenase/reductase family, featuring a conserved NAD+-binding domain required for its enzymatic activity. Expression of HSD3B1 is tissue-specific, predominantly localized in steroidogenic tissues such as the adrenal glands, gonads, and placenta. It is found in the smooth endoplasmic reticulum, where steroid biosynthesis occurs. HSD3B1 interacts with other enzymes in steroidogenic pathways, such as CYP17A1, facilitating androgen and estrogen synthesis. Genetic polymorphisms, such as the HSD3B1 (1245C) variant, have been associated with altered enzymatic activity, leading to increased androgen synthesis and resistance to androgen-deprivation therapy in prostate cancer. Dysregulation of HSD3B1 expression or activity is implicated in diseases such as metastatic castration-resistant prostate cancer, congenital adrenal hyperplasia, and hormone-related disorders. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref: Freitas, P.F.S., et al., Nat Rev Urol 22, 313 320)
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the internal region of human HSD3B1.

Application

Quality Control Testing

Evaluated by Western Blotting in Caco-2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected HSD3B1 in Caco-2 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HSD3B1 in human placenta tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected HSD3B1 in human placenta tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound with HSD3B1 peptide with at least one thousand-fold (1,000X) higher affinity than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 2M19 is a ZooMAb® rabbit recombinant monoclonal antibody that detects HSD3B1. It targets an epitope within 17 amino acids from the internal region.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.


Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

¿No ve la versión correcta?

Si necesita una versión concreta, puede buscar un certificado específico por el número de lote.

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico