Merck
  • Specific CP110 Phosphorylation Sites Mediate Anaphase Catastrophe after CDK2 Inhibition: Evidence for Cooperation with USP33 Knockdown.
Specific CP110 Phosphorylation Sites Mediate Anaphase Catastrophe after CDK2 Inhibition: Evidence for Cooperation with USP33 Knockdown.
Molecular cancer therapeutics (2015-08-26)
Shanhu Hu, Yun Lu, Bernardo Orr, Kristina Godek, Lisa Maria Mustachio, Masanori Kawakami, David Sekula, Duane A Compton, Sarah Freemantle, Ethan Dmitrovsky
RESUMEN
Chromosomal instability (CIN) is a hallmark of solid tumor biology and is implicated in carcinogenesis. Preferentially eliminating malignant cells by targeting CIN and aneuploidy is an attractive antineoplastic strategy. We previously reported that CDK2 antagonism causes lung cancer cells to undergo anaphase catastrophe and apoptosis through inhibition of phosphorylation of the centrosomal protein CP110. Cells with activating KRAS mutations were particularly sensitive to CDK2 inhibition due to downregulation of CP110 protein levels. This study investigated mechanisms of CDK2 antagonism that mediate anaphase catastrophe via changes in CP110 protein expression and how activated KRAS affects CP110 levels in lung cancers. Site-directed mutagenesis revealed candidate CDK phosphorylation sites of CP110 (residues Ser 170 and Thr 194) critical for conferring anaphase catastrophe by altering centrosome clustering in mitosis. Intriguingly, KRAS mutation can promote CP110 protein degradation by upregulating the ubiquitin ligase SCF(cyclinF), which targets CP110 protein for destabilization. Finally, CDK2 inhibitor response was enhanced when combined with knockdown of the deubiquitinase USP33 that in turn accelerates CP110 protein degradation. Thus, this study provides molecular pharmacologic insights into how CP110 expression regulates response to CDK2 inhibition. An improved understanding of in vitro antineoplastic mechanisms of combining CDK2 antagonism with induced CP110 repression provides a rationale for exploring clinical consequences of this strategy. Taken together, preclinical findings obtained from combining CDK2 inhibition with USP33 repression have implications for treating patients with non-small cell lung cancers.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Cicloheximida, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Cicloheximida, Biotechnology Performance Certified
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Metanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Metanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.