Merck
CREB expression mediates amyloid β-induced basal BDNF downregulation.
Neurobiology of aging (2015-05-31)
Elyse Rosa, Margaret Fahnestock
RESUMEN
In Alzheimer's disease, accumulation of amyloid-β (Aβ) is associated with loss of brain-derived neurotrophic factor (BDNF), synapses, and memory. Previous work demonstrated that Aβ decreases activity-induced BDNF transcription by regulating cyclic adenosine monophosphate response element binding protein (CREB) phosphorylation. However, the specific mechanism by which Aβ reduces basal BDNF expression remains unclear. Differentiated, unstimulated human neuroblastoma (SH-SY5Y) cells treated with oligomeric Aβ exhibited significantly reduced CREB messenger RNA compared with controls. Phosphorylated and total CREB proteins were decreased in both the cytoplasm and nucleus of Aβ-treated cells. However, neither pCREB129 nor pCREB133 levels were altered relative to total CREB levels. The protein kinase A activator forskolin increased pCREB133 levels and prevented Aβ-induced basal BDNF loss when administered before Aβ but did not rescue BDNF expression when administered later. These data demonstrate a new mechanism for Aβ-induced BDNF downregulation: in the absence of cell stimulation, Aβ downregulates basal BDNF levels via Aβ-induced CREB transcriptional downregulation, not changes in CREB phosphorylation. Thus, Aβ reduces basal and activity-induced BDNF expression by different mechanisms.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
L-Glutamina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Forskolina, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutamina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Forskolina, For use in molecular biology applications
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Adenina, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
SAFC
L-Glutamina
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Sigma-Aldrich
Adenina, BioReagent, suitable for cell culture