Direkt zum Inhalt
Merck

Fortfahren mit

GILF171503

Gilson™ PIPETMAN® DIAMOND Filter Tips

DF200ST TIPACK, volume 2-200 μL, sterile

Synonym(e):

micropipette filter tips

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41121600
eCl@ss:
32110208
Volume:
2-200 μL
Sterility:
sterile
Feature:
ATP free, DNA free, RNase and DNase free, endotoxin free, filter, graduated
Packaging:
pack of 960 EA (10 racks x96 tips)
Manufacturer/tradename:
Gilson F171503
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


material

conical polypropylene , yellow (color code)

description

calibrated: no

sterility

sterile

feature

ATP free, DNA free, RNase and DNase free, endotoxin free, filter, graduated

manufacturer/tradename

Gilson F171503

packaging

pack of 960 EA (10 racks x96 tips)

tip Overall L

51 mm

volume

2-200 μL

compatibility

Air Displacement Pipettes, Gilson MyPIPETMAN, PIPETMAN, PIPETMAN L, PIPETMAN M, PIPETMAN M Connected, PIPETMAN M96, PIPETMAX, PLATEMASTER

General description

PIPETMAN® DIAMOND Sterilized Filter Tips are designed to fit your PIPETMAN® perfectly and play a critical role in preventing aerosol-born contamination. They deliver accurate and precise pipetting when used in conjunction with Gilson™ pipettes. Compliant with Good Laboratory Practice (GLP).

Application

Biology, Biochemistry, immunoassays, Cell culture, Analytical chemistry

Features and Benefits

  • Accurate and Precise - Using PIPETMAN® DIAMOND Tips with your PIPETMAN® ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing. Volume graduations make it easy to visually check the volume aspirated.
  • Reliable - Gilson™ tips are manufactured in a clean room environment ensuring they are free of detectable contaminants that can affect your results. From mold to bench, our tips are guaranteed, certified, and traceable to meet the most stringent demands in many applications.
  • Convenient Packaging - Uniquely designed TIPACK™S feature a dual-purpose lid: hinged to easily open and close while using tips; or simply pull it out of the hinges to set on top or discard for high-throughput tip usage. TIPACK™S are compact, ergonomic, and eco-friendly.

Legal Information

Gilson is a trademark of Gilson, Inc.
Pipetman is a registered trademark of Gilson, Inc.
TIPACK is a trademark of Gilson, Inc.


Still not finding the right product?




Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen