Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
UNSPSC Code:
41106514
eCl@ss:
32160703
Biological source:
mouse embryo
Growth mode:
Adherent
Karyotype:
Not specified
Morphology:
Adherent monolayer of small spheroidal cells on gelatine
Manufacturer/tradename:
ECACC
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenProduktname
Mbd3KO, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
mouse embryo
manufacturer/tradename
ECACC
packaging
tube of 5 μg 11041301-DNA-5UG, pkg of vial of cells 11041301-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Adherent monolayer of small spheroidal cells on gelatine
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
-140 to -196°C
Biochem/physiol Actions
Mbd3KO stem cells have been genetically modified to produce a null phenotype for Mbd3, a critical determinant for lineage commitment in embryonic stem cells. This lack of Mbd3 (Mbd3 exons 2-7 have been replaced by an IRES-βgeo-pA sequence) allows them to be maintained in the absence of any exogenous factors and they fail to commit to developmental lineages. Upon re-introduction of the Mbd3 gene the cells are able to differentiate into identifiable bone, muscle, skin, fat and neuronal tissue. Mbd3 null embryonic stem cells can be used to study LIF-independent self-renewal to develop human stem cell lines and new cell culture media. They can also be used to develop specific stem cell lines for high throughput screening of therapeutic compounds. Finally they can be used to develop transgenics for the study of specific diseases or for the development of specific therapeutic stem cell lines for replacement therapies to treat certain diseases.
Mouse embryonic stem cell; Mbd3 knock-out
Preparation Note
GMEM (G5154) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442) + 0.1 mM 2-Mercaptoethanol (M6250) + 1000 Units/ml LIF. Use of LIF is optional, but its addition reduces differentiation.
Plastic ware is pre-coated with gelatine as follows: porcine gelatine (Sigma product number G1890) is dissolved in sterile water (0.5 g/500ml) at 50°C. The 0.1% solution is sterilized by filtration (0.22 μm). Add 0.1% gelatine to plastic ware to cover bottom, and incubate for 20 minutes at room temperature . Remove gelatine, wash with PBS once and replace with appropriate culture medium. The flask/dish must not be allowed to dry out. An ampoule of Mbd3KO cells is thawed in 37 °C water bath and the contents quickly transferred to a 15 ml centrifuge tube. Medium is added drop wise to 5 ml. Cells are centrifuged at 150 x g for 5 minutes. Cells are resuspended in 5 ml of medium. Cells should be plated at 4-5 x 104 cells/cm2 using gelatine coated flasks. Cells must be subcultured every 2-3 days using 0.25% trypsin or trypsin/EDTA. Colonies must not be allowed to touch each other as overgrowth will result in differentiation. Cultures must be incubated in a humidified 5% CO2/95% air incubator at 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.