Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Über diesen Artikel
Empirische Formel (Hill-System):
C18H18ClN3O2
CAS-Nummer:
Molekulargewicht:
343.81
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenagency
USP
API family
loxapine
packaging
pkg of 25 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
pharmaceutical
pharmaceutical
format
neat
storage temp.
−20°C
SMILES string
Clc1cc2c([o]c3c(nc2N4CC[N+](CC4)([O-])C)cccc3)cc1
InChI
1S/C18H18ClN3O2/c1-22(23)10-8-21(9-11-22)18-14-12-13(19)6-7-16(14)24-17-5-3-2-4-15(17)20-18/h2-7,12H,8-11H2,1H3
InChI key
QUSWANHLDLADDR-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Find answers to frequently asked questions on USP products in USP FAQs.
Application
Loxapine N-oxide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
Also, for use with USP monographs such as:
- Loxapine Succinate
- Loxapine Capsules
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Lagerklasse
13 - Non Combustible Solids
Was ist los?
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.