Direkt zum Inhalt
Merck

1A01110

USP

Dehydro Desloratadine

Pharmaceutical Analytical Impurity (PAI)

Synonym(e):

8-Chloro-11-(piperidin-4-ylidene)-11H-benzo[5,6]cyclohepta[1,2-b]pyridine, 13-Chloro-2-(piperidin-4-ylidene)-4-azatricyclo[9.4.0.0,3,8]pentadeca-1(11),3(8),4,6,9,12,14-heptaene

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern

Über diesen Artikel

Empirische Formel (Hill-System):
C19H17ClN2
CAS-Nummer:
Molekulargewicht:
308.80
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


grade

pharmaceutical analytical impurity (PAI)

agency

USP

API family

desloratadine

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Dehydro Desloratadine is a USP Pharmaceutical Analytical Impurity (PAI).
USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

Associated Drug Substance: Desloratadine
Therapeutic Area: Antihistamines

Application

Dehydro Desloratadine (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here


Lagerklasse

11 - Combustible Solids

Was ist los?

WGK 3

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

Die passende Version wird nicht angezeigt?

Wenn Sie eine bestimmte Version benötigen, können Sie anhand der Lot- oder Chargennummer nach einem spezifischen Zertifikat suchen.

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen