Produktname
1,2,2,6,6-Pentamethyl-4-piperidinol ≥99.0% (GC)
Produktnummer
03225
Produktmarke
sial
CAS-Nummer
2403-89-6
Molekulargewicht
171.28
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals or Flakes |
| PURITY (GC AREA %) | ≥ 99.0 |
| MELTING POINT | 72 - 76 C |
| WATER | ≤ 1.0 % |
| CARBON CONTENT | 70.12 % (theory) |
| HYDROGEN CONTENT | 12.36 % (theory) |
| NITROGEN CONTENT | 8.18 % (theory) |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |