Produktname
Acetyleugenol analytical standard
Produktnummer
04733
Produktmarke
sial
CAS-Nummer
93-28-7
Molekulargewicht
206.24
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless to Light Yellow |
| APPEARANCE (FORM) | Liquid |
| PURITY (GC AREA %) | ≥ 98.0 % |
| REFRACTIVE INDEX N20/D | 1.517 - 1.521 |
| WATER (COULOMETR.) | ≤ 0.3 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |