Produktname
Harmalin analytical standard
Produktnummer
08472
Produktmarke
sial
CAS-Nummer
304-21-2
Molekulargewicht
214.26
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow and Faint Beige to Light Beige and Faint Brown to Light Brown |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 95.0 % |
| CARBON CONTENT | 72.87 % (Theory) |
| HYDROGEN CONTENT | 6.59 % (Theory) |
| NITROGEN CONTENT | 13.07 % (Theory) |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |