Produktname
Thiophenol analytical standard
Produktnummer
09286
Produktmarke
sial
CAS-Nummer
108-98-5
Molekulargewicht
110.18
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| PURITY (GC AREA %) | ≥ 99.5 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 24 MONTHS |