Produktname
Cloransulam-methyl PESTANAL®, analytical standard
Produktnummer
32147
Produktmarke
sial
CAS-Nummer
147150-35-4
Molekulargewicht
429.81
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| MELTING POINT | 204 - 210 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |