Produktname
Desogestrel VETRANAL®, analytical standard
Produktnummer
32809
Produktmarke
sial
CAS-Nummer
54024-22-5
Molekulargewicht
310.47
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| MELTING POINT | 104 - 112 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |