Produktname
Ametrin PESTANAL®, analytical standard
Produktnummer
45321
Produktmarke
sial
CAS-Nummer
834-12-8
Molekulargewicht
227.33
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (HPLC AREA %) | ≥ 98.0 % |
| MELTING POINT | 85 - 94 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |