Produktname
Chlorthaldimethyl PESTANAL®, analytical standard
Produktnummer
45397
Produktmarke
sial
CAS-Nummer
1861-32-1
Molekulargewicht
331.96
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Light Yellow and Faint Beige to Light Beige and Faint Brown to Light Brown |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 98.0 % |
| MELTING POINT | 155 - 161 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |