Produktname
1,2,3,4-Tetrachlorbenzol PESTANAL®, analytical standard
Produktnummer
46014
Produktmarke
sial
CAS-Nummer
634-66-2
Molekulargewicht
215.89
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 98.0 % |
| MELTING POINT | 44 - 50 C |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |