Produktname
Methamidophos -Lösung 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Produktnummer
46342
Produktmarke
sial
CAS-Nummer
10265-92-6
Molekulargewicht
141.13
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| CONCENTRATION COMP1 | ~ 100 ug/ml |
| SOLVENT | ACETONITRILE |
| PURITY (GC AREA %) | ≥ 95.0 % |
| IDENTIFICATION (GC-MS) | CORRESPONDS TO REQUIREMENTS |
| REFRACTIVE INDEX N20/D | 1.3420 - 1.3460 |
| EXPIRY PERIOD | 36 MONTHS |